1-Chloro-1,1,2-trifluoro-2-iodoethane 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2-iodo-1,1,2-trifluoroethane
PC1934-25G PC1934-5G PC1934-100G
APOSPC1934-25GEA 271 EUR
APOSPC1934-25G APOSPC1934-5G APOSPC1934-100G
1-Chloro-1,1,2-trifluoro-2-iodoethane 97%
1-Chloro-1,1,2-trifluoro-2-iodoethane
Formula: C₂HClF₃I
MW: 244.38 g/mol
Boiling Pt: 77 °C/630mmHg
Density: 2.181 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042138
CAS Number: 354-26-7
EINECS: 206-553-6