Tetrabutylammonium fluoride 1.0 M THF
Supplier: Apollo Scientific
Danger
PC1945-1L PC1945-250ML
APOSPC1945-1LEA 248 EUR
APOSPC1945-1L APOSPC1945-250ML
Tetrabutylammonium fluoride 1.0 M THF
Tetrabutylammonium fluoride
Formula: C₁₆H₃₆FN
MW: 261.47 g/mol
Boiling Pt: 65…67 °C
Density: 0.903 g/cm³ (25 °C)
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011747
CAS Number: 429-41-4
EINECS: 207-057-2
UN: 1993
ADR: 3 III