4-Chloro-3-nitrobenzotrifluoride 98%
Supplier: Apollo Scientific
Synonyms: 1-Chloro-2-nitro-4-(trifluoromethyl)benzene, 4-Chloro-α-α-α-trifluoro-3-nitrotoluene, 4-Chloro-3-nitrobenzotrifluoride
PC1950-1KG PC1950-250G PC1950-5KG
APOSPC1950-1KGEA 36.4 EUR
APOSPC1950-1KG APOSPC1950-250G APOSPC1950-5KG
4-Chloro-3-nitrobenzotrifluoride 98%
4-Chloro-3-nitrobenzotrifluoride
Formula: C₇H₃ClF₃NO₂
Boiling Pt: 220…223 °C
Melting Pt: –2 °C
Density: 1.511 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007084
CAS Number: 121-17-5
UN: 2307
ADR: 6.1,II