2-Amino-3-fluoropyridine 98%
Supplier: Apollo Scientific
Warning
PC1957-25G PC1957-5G PC1957-100G
APOSPC1957-25GEA 114 EUR
APOSPC1957-25G APOSPC1957-5G APOSPC1957-100G
2-Amino-3-fluoropyridine 98%
2-Amino-3-fluoropyridine
Formula: C₅H₅FN₂
MW: 112.11 g/mol
Melting Pt: 39…45 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD04114135
CAS Number: 21717-95-3