5-Chloro-2-nitrobenzotrifluoride 99%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-1-nitro-2-(trifluoromethyl)benzene, 5-Chloro-2-nitrobenzotrifluoride, 1-Chloro-4-nitro-3-(trifluoromethyl)benzene, 5-Chloro-α,α,α-trifluoro-2-nitrotoluene
PC1960-100G PC1960-25G
APOSPC1960-100GEA 93.1 EUR
APOSPC1960-100G APOSPC1960-25G
5-Chloro-2-nitrobenzotrifluoride 99%
5-Chloro-2-nitrobenzotrifluoride
Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 222…224 °C
Melting Pt: 21…22 °C
Density: 1.526 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00007298
CAS Number: 118-83-2
UN: 3082
ADR: 9+EHS III