2-Amino-5-chloro-3-fluoropyridine 95%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Chloro-3-fluoro-2-pyridylamine, 5-Chloro-3-fluoropyridin-2-amine
PC1964-25G PC1964-5G PC1964-100G
APOSPC1964-25GEA 59.6 EUR
APOSPC1964-25G APOSPC1964-5G APOSPC1964-100G
2-Amino-5-chloro-3-fluoropyridine 95%
2-Amino-5-chloro-3-fluoropyridine
Formula: C₅H₄ClFN₂
MW: 146.55 g/mol
Melting Pt: 91…95 °C
Storage Temperature: Ambient
MDL Number: MFCD09878434
CAS Number: 246847-98-3
UN: 2811
ADR: 6.1 III