8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyridin-2-amine
Supplier: Apollo Scientific
PC200001-1G PC200001-500MG
APOSPC200001-1GEA 210 EUR
APOSPC200001-1G APOSPC200001-500MG
8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyridin-2-amine
8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formula: C₇H₄ClF₃N₄
MW: 236.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22383935
CAS Number: 1206640-61-0