Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methyl]amino}benzoate
Supplier: Apollo Scientific
PC200022-1G PC200022-500MG
APOSPC200022-1GEA 210 EUR
APOSPC200022-1G APOSPC200022-500MG
Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methyl]amino}benzoate
Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methyl]amino}benzoate
Formula: C₁₅H₁₄FNO₃
MW: 275.28 g/mol
Melting Pt: 130…131 °C
Storage Temperature: Ambient
MDL Number: MFCD22573519
CAS Number: 1407522-00-2