Methyl-5-(trifluoromethyl)-1-benzofuran-2-carboxylate 95+%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-(trifluoromethyl)benzofuran-2-carboxylate
PC200023-100MG PC200023-250MG
APOSPC200023-100MGEA 82.8 EUR
APOSPC200023-100MG APOSPC200023-250MG
Methyl-5-(trifluoromethyl)-1-benzofuran-2-carboxylate 95+%
Methyl-5-(trifluoromethyl)-1-benzofuran-2-carboxylate
Formula: C₁₁H₇F₃O₃
MW: 244.17 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD22573538
CAS Number: 1407521-96-3