6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one
Supplier: Apollo Scientific
Warning
Synonyms: 6,7-Dichloro-3-(trifluoromethyl)quinoxalin-2(1H)-one, 6,7-Dichloro-2-hydroxy-3-(trifluoromethyl)quinoxaline
PC200039-1G PC200039-500MG PC200039-5G
APOSPC200039-1GEA 201 EUR
APOSPC200039-1G APOSPC200039-500MG APOSPC200039-5G
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one
Formula: C₉H₃Cl₂F₃N₂O
MW: 283.300 g/mol
Melting Pt: 242…244 °C
Storage Temperature: Ambient
MDL Number: MFCD09801051
CAS Number: 477857-25-3