3-(Trifluoromethyl)-1H-1,2,4-triazole 97%
Supplier: Apollo Scientific
Warning
PC200058-1G PC200058-5G PC200058-250MG
APOSPC200058-1GEA 88.6 EUR
APOSPC200058-1G APOSPC200058-5G APOSPC200058-250MG
3-(Trifluoromethyl)-1H-1,2,4-triazole 97%
3-(Trifluoromethyl)-1H-1,2,4-triazole
Formula: C₃H₂F₃N₃
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD11847494
CAS Number: 60406-75-9