tert-Butyl-4-(((trifluoromethyl)sulphonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 4-(((trifluoromethyl)sulphonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate, 1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridin-4-yl trifluoromethanesulphonate
PC200076-25G PC200076-5G
APOSPC200076-25GEA 362 EUR
APOSPC200076-25G APOSPC200076-5G
tert-Butyl-4-(((trifluoromethyl)sulphonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate
tert-Butyl-4-(((trifluoromethyl)sulphonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate
Formula: C₁₁H₁₆F₃NO₅S
MW: 331.3087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997858
CAS Number: 138647-49-1