2-Trifluoromethyl-quinazolin-4-ol
Supplier: Apollo Scientific
Synonyms: 2-(Trifluoromethyl)quinazolin-4(3H)-one
PC200079-1G PC200079-500MG PC200079-5G
APOSPC200079-1GEA 311 EUR
APOSPC200079-1G APOSPC200079-500MG APOSPC200079-5G
2-Trifluoromethyl-quinazolin-4-ol
2-Trifluoromethyl-quinazolin-4-ol
Formula: C₉H₅F₃N₂O
MW: 214.15 g/mol
Melting Pt: 248…251 °C
Storage Temperature: Ambient
MDL Number: MFCD01764613
CAS Number: 26059-81-4