2,3-Dichloro-5-(difluoromethyl)pyridine
Supplier: Apollo Scientific
PC200125-1G PC200125-500MG
APOSPC200125-1GEA 364 EUR
APOSPC200125-1G APOSPC200125-500MG
2,3-Dichloro-5-(difluoromethyl)pyridine
2,3-Dichloro-5-(difluoromethyl)pyridine
Formula: C₆H₃Cl₂F₂N
MW: 198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22199295
CAS Number: 71690-06-7