5-Bromo-6-(trifluoromethyl)-4-pyrimidinol
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-6-(trifluoromethyl)pyrimidin-4-ol, 5-Bromo-4-hydroxy-6-(trifluoromethyl)pyrimidine
PC200154-1G PC200154-5G PC200154-10G
APOSPC200154-1GEA 29 EUR
APOSPC200154-1G APOSPC200154-5G APOSPC200154-10G
5-Bromo-6-(trifluoromethyl)-4-pyrimidinol
5-Bromo-6-(trifluoromethyl)-4-pyrimidinol
Formula: C₅H₂BrF₃N₂O
MW: 242.99 g/mol
Melting Pt: 225…227 °C
Storage Temperature: Ambient
MDL Number: MFCD09901776
CAS Number: 942060-14-2