Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 3-(Trifluoromethyl)pyrazole-5-carboxylate
PC200156-1G
APOSPC200156-1GEA 89.9 EUR
APOSPC200156-1G
Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate
Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate
Formula: C₇H₇F₃N₂O₂
MW: 208.14 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD04111238
CAS Number: 129768-30-5