Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
PC200165-1G PC200165-250MG PC200165-5G
APOSPC200165-1GEA 109 EUR
APOSPC200165-1G APOSPC200165-250MG APOSPC200165-5G
Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₉F₃N₂O₂
MW: 258.20 g/mol
Melting Pt: 125…126 °C
Storage Temperature: Ambient
MDL Number: MFCD11100212
CAS Number: 860457-99-4