Ethyl-8-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Warning
PC200167-1G PC200167-500MG
APOSPC200167-1GEA 192 EUR
APOSPC200167-1G APOSPC200167-500MG
Ethyl-8-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 8-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₁H₈BrF₃N₂O₂
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD12025881
CAS Number: 1038389-86-4