Ethyl 2-[(tert-butoxycarbonyl)amino]-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate
Supplier: Apollo Scientific
Synonyms: Ethyl 2-amino-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate, 2-BOC protected
PC200171-1G PC200171-500MG
APOSPC200171-1GEA 210 EUR
APOSPC200171-1G APOSPC200171-500MG
Ethyl 2-[(tert-butoxycarbonyl)amino]-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate
Ethyl-2-[(tert-butoxycarbonyl)amino]-4-(trifluoromethyl)-1,3-oxazole-5-carboxylate
Formula: C₁₂H₁₅F₃N₂O₅
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD12827772
CAS Number: 1186405-12-8