5-Methoxy-2-(trifluoromethyl)-4-pyrimidinol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-5-methoxy-2-(trifluoromethyl)pyrimidine
PC200188-1G PC200188-500MG
APOSPC200188-1GEA 452 EUR
APOSPC200188-1G APOSPC200188-500MG
5-Methoxy-2-(trifluoromethyl)-4-pyrimidinol 95%
5-Methoxy-2-(trifluoromethyl)-4-pyrimidinol
Formula: C₆H₅F₃N₂O₂
MW: 194.11 g/mol
Melting Pt: 225…227 °C
Storage Temperature: Ambient
MDL Number: MFCD11100214
CAS Number: 808770-42-5