6-Fluoroimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
PC200221-1G PC200221-10G PC200221-5G
APOSPC200221-1GEA 79.9 EUR
APOSPC200221-1G APOSPC200221-10G APOSPC200221-5G
6-Fluoroimidazo[1,2-a]pyridine
6-Fluoroimidazo[1,2-a]pyridine
Formula: C₇H₅FN₂
MW: 136.1300 g/mol
Melting Pt: 70…71 °C
Storage Temperature: Ambient
MDL Number: MFCD09994632
CAS Number: 139022-27-8