8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decan-3-one
Supplier: Apollo Scientific
PC200237-1G PC200237-500MG
APOSPC200237-1GEA 190 EUR
APOSPC200237-1G APOSPC200237-500MG
8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decan-3-one
8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decan-3-one
Formula: C₁₃H₁₃ClF₃N₃OS
MW: 351.78 g/mol
Melting Pt: 206…209 °C
Storage Temperature: Ambient
MDL Number: MFCD17167322
CAS Number: 1246738-29-3