4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
Supplier: Apollo Scientific
PC200241-1G PC200241-500MG PC200241-5G
APOSPC200241-1GEA 336 EUR
APOSPC200241-1G APOSPC200241-500MG APOSPC200241-5G
4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
Formula: C₁₃H₈F₃NO
MW: 251.21 g/mol
Melting Pt: 67…68 °C
Storage Temperature: Ambient
MDL Number: MFCD10758064
CAS Number: 847446-85-9