2,3-Dichloro-5-(trifluoromethyl)isonicotinamide
Supplier: Apollo Scientific
PC200242-1G PC200242-500MG
APOSPC200242-1GEA 415 EUR
APOSPC200242-1G APOSPC200242-500MG
2,3-Dichloro-5-(trifluoromethyl)isonicotinamide
2,3-Dichloro-5-(trifluoromethyl)isonicotinamide
Formula: C₇H₃Cl₂F₃N₂O
Melting Pt: 185…186 °C
Storage Temperature: Ambient
MDL Number: MFCD14581646
CAS Number: 1198475-46-5