Ethyl-3-amino-1,6-dimethyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Supplier: Apollo Scientific
PC200247-1G PC200247-500MG
APOSPC200247-1GEA 415 EUR
APOSPC200247-1G APOSPC200247-500MG
Ethyl-3-amino-1,6-dimethyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Ethyl-3-amino-1,6-dimethyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Formula: C₁₃H₁₄F₃N₃O₂
MW: 301.27 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD20921605
CAS Number: 1353877-98-1