8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decane hydrochloride
Supplier: Apollo Scientific
PC200261-1G PC200261-500MG
APOSPC200261-1GEA 248 EUR
APOSPC200261-1G APOSPC200261-500MG
8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decane hydrochloride
8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decane hydrochloride
Formula: C₁₃H₁₆Cl₂F₃N₃S
MW: 374.26 g/mol
Melting Pt: 175…176 °C
Storage Temperature: Ambient
MDL Number: MFCD16890142
CAS Number: 1242267-83-9