1-[(4-Chlorophenyl)carbonyl]-3-[5-(trifluoromethyl)pyridin-2-yl]thiourea 95%
Supplier: Apollo Scientific
PC200262-1G PC200262-500MG
APOSPC200262-1GEA 306 EUR
APOSPC200262-1G APOSPC200262-500MG
1-[(4-Chlorophenyl)carbonyl]-3-[5-(trifluoromethyl)pyridin-2-yl]thiourea 95%
1-[(4-Chlorophenyl)carbonyl]-3-[5-(trifluoromethyl)pyridin-2-yl]thiourea
Formula: C₁₄H₉ClF₃N₃OS
Storage Temperature: Ambient
MDL Number: MFCD20921588
CAS Number: 1353878-23-5