Methyl-2,6-dioxo-5-(trifluoromethyl)-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate
Supplier: Apollo Scientific
PC200282-1G PC200282-500MG
APOSPC200282-1GEA 633 EUR
APOSPC200282-1G APOSPC200282-500MG
Methyl-2,6-dioxo-5-(trifluoromethyl)-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate
Methyl-2,6-dioxo-5-(trifluoromethyl)-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate
Formula: C₇H₅F₃N₂O₄
Melting Pt: 225…226 °C
Storage Temperature: Ambient
MDL Number: MFCD12922685
CAS Number: 936476-63-0