6-Cyclopropyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine
Supplier: Apollo Scientific
Synonyms: 3-Amino-6-cyclopropyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine
PC200284-1G PC200284-500MG
APOSPC200284-1GEA 519 EUR
APOSPC200284-1G APOSPC200284-500MG
6-Cyclopropyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine
6-Cyclopropyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine
Formula: C₁₀H₉F₃N₄
MW: 242.21 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD16890144
CAS Number: 1242267-90-8