Methyl 5-(trifluoromethoxy)-1H-indazole-3-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-(trifluoromethoxy)-1H-indazole-3-carboxylate
PC200315-500MG PC200315-250MG PC200315-1G
APOSPC200315-500MGEA 190 EUR
APOSPC200315-500MG APOSPC200315-250MG APOSPC200315-1G
Methyl 5-(trifluoromethoxy)-1H-indazole-3-carboxylate
Methyl-5-(trifluoromethoxy)-1H-indazole-3-carboxylate
Formula: C₁₀H₇F₃N₂O₃
Melting Pt: 181…182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12498703
CAS Number: 932041-12-8