Ethyl-4-chloro-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Warning
PC200338-1G PC200338-500MG
APOSPC200338-1GEA 431 EUR
APOSPC200338-1G APOSPC200338-500MG
Ethyl-4-chloro-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Ethyl-4-chloro-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Formula: C₇H₆ClF₃N₂O₂
MW: 242.59 g/mol
Melting Pt: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD22373662
CAS Number: 855345-12-9