3-Fluoro-5-formylbenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC200339-5G PC200339-25G
APOSPC200339-5GEA 75.4 EUR
APOSPC200339-5G APOSPC200339-25G
3-Fluoro-5-formylbenzonitrile 98%
3-Fluoro-5-formylbenzonitrile
Formula: C8H4FNO
MW: 149.1200 g/mol
Boiling Pt: 205 °C
Melting Pt: 82…84 °C
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD04974123
CAS Number: 1003708-42-6