6-Fluoroisoquinoline 95+%

Supplier: Apollo Scientific
Warning

PC201019-1G PC201019-5G
APOSPC201019-1GEA 55.1 EUR
APOSPC201019-1G APOSPC201019-5G
6-Fluoroisoquinoline 95+%
6-Fluoroisoquinoline
Formula: C₉H₆FN
MW: 147.1491 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226829
CAS Number: 1075-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR