3-Fluoro-4-cyanopyridine 98%
Supplier: Apollo Scientific
PC201025-1G PC201025-25G PC201025-5G
APOSPC201025-1GEA 69.8 EUR
APOSPC201025-1G APOSPC201025-25G APOSPC201025-5G
3-Fluoro-4-cyanopyridine 98%
3-Fluoro-4-cyanopyridine
Formula: C6H3FN2
MW: 122.10 g/mol
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD04112524
CAS Number: 113770-88-0
UN: 3439
ADR: 6.1,III