4-Bromo-2-fluoro-3-formylpyridine 97%
Supplier: Apollo Scientific
Synonyms: 4-Bromo-2-fluoronicotinaldehyde
PC201111-1G PC201111-250MG PC201111-5G
APOSPC201111-1GEA 30.7 EUR
APOSPC201111-1G APOSPC201111-250MG APOSPC201111-5G
4-Bromo-2-fluoro-3-formylpyridine 97%
4-Bromo-2-fluoro-3-formylpyridine
Formula: C₆H₃BrFNO
MW: 204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09870050
CAS Number: 128071-77-2