Methyl-4-amino-2,6-difluorobenzoate 98%
Supplier: Apollo Scientific
Synonyms: Methyl 4-amino-2,6-difluorobenzoate
PC201187-100MG PC201187-1G PC201187-250MG
APOSPC201187-100MGEA 101 EUR
APOSPC201187-100MG APOSPC201187-1G APOSPC201187-250MG
Methyl-4-amino-2,6-difluorobenzoate 98%
Methyl-4-amino-2,6-difluorobenzoate
Formula: C₈H₇F₂NO₂
MW: 187.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16038807
CAS Number: 191478-99-6