3,5-Difluoro-4-hydroxybenzonitrile 96%
Supplier: Apollo Scientific
PC201222-10G PC201222-5G
APOSPC201222-10GEA 303 EUR
APOSPC201222-10G APOSPC201222-5G
3,5-Difluoro-4-hydroxybenzonitrile 96%
3,5-Difluoro-4-hydroxybenzonitrile
Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033181
CAS Number: 2967-54-6