2-Chloro-1,5-difluoro-3-nitrobenzene 97%
Supplier: Apollo Scientific
PC201239-1G PC201239-250MG
APOSPC201239-1GEA 201 EUR
APOSPC201239-1G APOSPC201239-250MG
2-Chloro-1,5-difluoro-3-nitrobenzene 97%
2-Chloro-1,5-difluoro-3-nitrobenzene
Formula: C₆H₂ClF₂NO₂
MW: 193.54 g/mol
Storage Temperature: Ambient
CAS Number: 36556-59-9