5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde 98%
Supplier: Apollo Scientific
Synonyms: 5-(2-Fluorophenyl)-1H-pyrrole-3-carbaldehyde
PC201345-25G
APOSPC201345-25GEA 112 EUR
APOSPC201345-25G
5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde 98%
5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde
Formula: C₁₁H₈FNO
MW: 189.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11875867
CAS Number: 881674-56-2