4-Chloro-5-fluoro-2-methylpyrimidine 96%
Supplier: Apollo Scientific
PC201366-250MG
PC201366-1G
PC201366-5G
APOSPC201366-250MGEA
69.6
EUR
APOSPC201366-250MG
APOSPC201366-1G
APOSPC201366-5G
4-Chloro-5-fluoro-2-methylpyrimidine 96%
4-Chloro-5-fluoro-2-methylpyrimidine
Formula:
C₅H₄ClFN₂ MW: 146.5501 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09834964 CAS Number: 898044-50-3 |