3-Fluoro-N-methylbenzylamine 96%
Supplier: Apollo Scientific
Danger
Synonyms: (3-Fluorobenzyl)methylamine, 1-(3-Fluorophenyl)-N-methylmethanamine
PC201369-25G
APOSPC201369-25GEA 148 EUR
APOSPC201369-25G
3-Fluoro-N-methylbenzylamine 96%
3-Fluoro-N-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 183…184 °C
Density: 1.015 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04623547
CAS Number: 90389-84-7