1,3-Difluoro-5-methyl-2-nitrobenzene 98%
Supplier: Apollo Scientific
PC201377-250MG PC201377-1G PC201377-5G
APOSPC201377-250MGEA 91.5 EUR
APOSPC201377-250MG APOSPC201377-1G APOSPC201377-5G
1,3-Difluoro-5-methyl-2-nitrobenzene 98%
1,3-Difluoro-5-methyl-2-nitrobenzene
Formula: C₇H₅F₂NO₂
MW: 173.1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527695
CAS Number: 932373-92-7