4-Fluoropyridin-2(1H)-one 98%
Supplier: Apollo Scientific
Warning
PC201387-250MG PC201387-1G
APOSPC201387-250MGEA 29 EUR
APOSPC201387-250MG APOSPC201387-1G
4-Fluoropyridin-2(1H)-one 98%
4-Fluoropyridin-2(1H)-one
Formula: C₅H₄FNO
MW: 113.090 g/mol
Melting Pt: 174…175 °C
Storage Temperature: Ambient
MDL Number: MFCD07437952
CAS Number: 96530-75-5