5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride 97%
Supplier: Apollo Scientific
Danger
PC2021-1G PC2021-250MG
APOSPC2021-1GEA 75.4 EUR
APOSPC2021-1G APOSPC2021-250MG
5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride 97%
5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride
Formula: C₁₂H₅Cl₂F₃O₂
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00275554
CAS Number: 175276-61-6
UN: 1759
ADR: 8,III