2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid 95%
Supplier: Apollo Scientific
Warning
PC2024-1G PC2024-250MG
APOSPC2024-1GEA 62.5 EUR
APOSPC2024-1G APOSPC2024-250MG
2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid 95%
2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid
Formula: C₁₁H₆ClF₃N₂O₂
MW: 290.63 g/mol
Melting Pt: 137…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00068142
CAS Number: 98534-80-6