3-(3-Fluorophenyl)propionic acid 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(3-Fluorophenyl)propanoic acid
PC2030-250MG PC2030-5G PC2030-10G
APOSPC2030-250MGEA 14.7 EUR
APOSPC2030-250MG APOSPC2030-5G APOSPC2030-10G
3-(3-Fluorophenyl)propionic acid 98%
3-(3-Fluorophenyl)propionic acid
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 125 °C/10.5mmHg
Melting Pt: 43…47 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01310821
CAS Number: 458-45-7