1-Chloro-2,3,3-trifluorocyclobutene 97%
Supplier: Apollo Scientific
Danger
PC2035-1G PC2035-5G
APOSPC2035-1GEA 87 EUR
APOSPC2035-1G APOSPC2035-5G
1-Chloro-2,3,3-trifluorocyclobutene 97%
1-Chloro-2,3,3-trifluorocyclobutene
Formula: C₄H₂ClF₃
MW: 142.51 g/mol
Boiling Pt: 51…52 °C
Density: 1.355 g/cm³ (25 °C)
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00041524
CAS Number: 694-62-2
UN: 1993
ADR: 3,II