2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Chloro-4-(trifluoromethyl)benzene-1-sulphonyl chloride
PC2073-1G PC2073-25G PC2073-5G
APOSPC2073-1GEA 21.9 EUR
APOSPC2073-1G APOSPC2073-25G APOSPC2073-5G
2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride 97%
2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃Cl₂F₃O₂S
MW: 279.07
Boiling Pt: 70…71 °C/0.5mmHg
Melting Pt: 37…39 °C
Density: 1.597 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00052912
CAS Number: 175205-54-6
UN: 3265
ADR: 8 III