2-Chloro-5-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-5-trifluoromethylpyridine
PC2075B-100G PC2075B-25G
APOSPC2075B-100GEA 42.2 EUR
APOSPC2075B-100G APOSPC2075B-25G
2-Chloro-5-(trifluoromethyl)pyridine 97%
2-Chloro-5-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 152…153 °C
Melting Pt: 29…33 °C
Density: 1.417 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00042225
CAS Number: 52334-81-3
EINECS: 257-856-5