4-Chloro-2-(trifluoromethyl)quinoline 97%
Supplier: Apollo Scientific
Warning
PC2075K-1G PC2075K-5G
APOSPC2075K-1GEA 65.5 EUR
APOSPC2075K-1G APOSPC2075K-5G
4-Chloro-2-(trifluoromethyl)quinoline 97%
4-Chloro-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MW: 231.61 g/mol
Melting Pt: 34…38 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00153105
CAS Number: 1701-24-2